MA VPH surrogate standard (2,5-dibromotoluene)
Catalog # REST30435
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml in P&T methanol
- Pk:1 mL
- CAS No.:615-59-8
- Description:MA VPH surrogate standard (2,5-Dibromotoluene)
- Environmentally Preferable:
- Formula:C₇H₆Br₂
- Description:2,5-Dibromotoluene
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.