Order Entry
Belgium
ContactUsLinkComponent
Volatiles MegaMix standard with gases
Volatiles MegaMix standard with gases
Catalog # REST30603
Supplier:  Restek
CAS Number:  
undefined
Volatiles MegaMix standard with gases
Catalog # REST30603
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 µg/ml each in P&T methanol
  • Pk:
    1 Ampoul
  • Description:
    Volatiles MegaMix with gases
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

60 components

The volatiles megamix standard with gases consists of 60 components they are benzene, bromobenzene, bromochloromethane, bromodichloromethane, bromoform, bromomethane , n-butylbenzene, sec-butylbenzene, tert-butylbenzene, carbon tetrachloride, chloroform, chloromethane , 2-chlorotoluene, 4-chlorotoluene, dibromochloromethane, 1,2-dibromo-3-chloropropane , 1,2-dibromoethane , dibromomethane, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, dichlorodifluoromethane , 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, 1,3-dichloropropane, 2,2-dichloropropane, 1,1-dichloropropene, cis-1,3-dichloropropene, trans-1,3-dichloropropene, ethylbenzene, hexachloro-1,3-butadiene , isopropylbenzene , 4-isopropyltoluene , methylene chloride , naphthalene, n-propylbenzene, styrene, 1,1,1,2-tetrachloroethane, 1,1,2,2-tetrachloroethane, tetrachloroethene, toluene, 1,2,3-trichlorobenzene, 1,2,4-trichlorobenzene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, trichlorofluoromethane , 1,2,3-trichloropropane, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, vinyl chloride, m-xylene, o-xylene, p-xylene .

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.