Order Entry
Belgium
ContactUsLinkComponent
Calibration check mix, 8240/8260 (6 components)
Calibration check mix, 8240/8260 (6 components)
Catalog # REST30427
Supplier:  Restek
CAS Number:  
undefined
Calibration check mix, 8240/8260 (6 components)
Catalog # REST30427
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    Calibration check mix, 8240/8260
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains chloroform, 1,1-dichloroethene, 1,2-dichloropropane, ethylbenzene, toluene, vinyl chloride.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.