Order Entry
Belgium
ContactUsLinkComponent
Calibration mix, 8260 MegaMix (76 components)
Calibration mix, 8260 MegaMix (76 components)
Catalog # REST30633
Supplier:  Restek
CAS Number:  
undefined
Calibration mix, 8260 MegaMix (76 components)
Catalog # REST30633
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P&T methanol
  • Pk:
    1 mL
  • Description:
    MegaMix calibration mix 8260
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This calibration mix contains 76 compounds in one convenient ampoule for the analysis of VOCs in solid waste.

  • Quantitatively analysed to confirm concentration and identity of each compound

Analyze VOCs in solid waste by EPA Method 8260 using our popular MegaMix calibration mix. For an identical mix that does not contain pentachloroethane, see our revised mix (cat.# REST30152).

Certifications: CRM.