Order Entry
Belgium
ContactUsLinkComponent
GC multi-residue pesticide standard #9 (8 components)
GC multi-residue pesticide standard #9 (8 components)
Catalog # REST32571
Supplier:  Restek
CAS Number:  
undefined
GC multi-residue pesticide standard #9 (8 components)
Catalog # REST32571
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #9, 8 components, 100 μg/ml each in toluene
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The multiresidue accurately identify and quantify pesticide residues by gc-ms/ms in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability

Ampul 7 of 9 in the GC-MS/MS multiresidue pesticide standard kit.

The multiresidue pesticide standard consists of eight components disulphoton, fonofos, methyl parathion, mevinphos, parathion (ethyl parathion), phorate, piperonyl butoxide, triazophos.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Ordering information: NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.