Orders LinkContactUsLinkComponent
Laboratory software, Labworldsoft 6
Supplier:   IKA
Laboratory software, Labworldsoft 6
Supplier:  IKA
Certificates
Laboratory software, Labworldsoft 6
Laboratory software, Labworldsoft 6
3 Options Available Below
Product Details & Documents
Labworldsoft 6 is a laboratory software to control lab devices, automate processes, and to document your measurements and results. The software provides access to a huge library of different laboratory devices.
  • Easy recording of various physical parameters
  • Function modules to create time triggered or event triggered configurations
  • Communication through RS232, USB, Bluetooth or network
  • Supports formats .csv, .xslx and ASCII
  • Compatibility with Windows 7, 8 and 10
Labworldsoft does not integrate only IKA laboratory devices, but supports several other device from different manufacturers as well. You stay flexible in the choice of your lab devices you want to use and profit from the whole device range of the laboratory world.
Delivery information: Includes one year of support with new versions of Labworldsoft 6.
Product Family Options
Product Information
  • For
    Availability
    Price
  • IKAA20019397
    Labworldsoft® 6 Starter
    Labworldsoft® 6 Starter
    Small configurations with up to 3 devices can be controlled at the same time
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 500,00 €
    6 500,00 €
    1 * 1 ST
       6 500,00 €
  • IKAA20017366
    Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    All configurations with any number of devices can be controlled at the same time
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14 820,00 €
    14 820,00 €
    1 * 1 ST
       14 820,00 €
  • IKAA20101872
    Labworldsoft® 6 Visc
    Labworldsoft® 6 Visc
    Viscosity measurements for thermostats and pH meters of all manufacturers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 450,00 €
    1 450,00 €
    1 * 1 ST
       1 450,00 €