Order Entry
Belgium
ContactUsLinkComponent
DHA Naphthenes standard (26 components)
DHA Naphthenes standard (26 components)
Catalog # REST30728
Supplier:  Restek
CAS Number:  
undefined
DHA Naphthenes standard (26 components)
Catalog # REST30728
Supplier:  Restek
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1
  • Description:
    DHA naphthenes standard

Specifications

About this item

This naphthenes standard is produced for refineries performing detailed hydrocarbon analysis (DHA) of crude oil feedstocks and fuels.

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for qualitative calibration.

Concentration: Neat, 0,15 ml in autosampler vial.