ContactUsLinkComponent
FAME #5 mix
FAME #5 mix
Catalog # REST35014
Supplier:  Restek
CAS Number:  
undefined
FAME #5 mix
Catalog # REST35014
Supplier:  Restek
CAS Number:  
Specifications
  • Pack type:
    Ampoule
  • Pk:
    1
  • Description:
    FAME #5, each mixture has minimum 50 mg/ampoule
Specifications
About this item
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C16 - C24)
This mix contains five components such as C16:01 methyl palmitoleate (20%), C18:01 methyl oleate (20%), C20:01 methyl eicosenoate (20%), C22:01 methyl erucate (20%), C24:01:00 methyl nervonate (20%).