Order Entry
Belgium
ContactUsLinkComponent
Human ROMO1 (reactive oxygen species modulator 1) ELISA kit, Immunotag™
Human ROMO1 (reactive oxygen species modulator 1) ELISA kit, Immunotag™
Catalog # GENOIT3170
Supplier:  G-Biosciences
CAS Number:  
Human ROMO1 (reactive oxygen species modulator 1) ELISA kit, Immunotag™
Catalog # GENOIT3170
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Immunotag™ human ROMO1 (reactive oxygen species modulator 1) ELISA kit
  • Detection range:
    31.2 - 2000 pg/ml
  • Sensitivity:
    18.75 pg/ml

Specifications

About this item

Immunotag™ Human ROMO1 (Reactive oxygen species modulator 1) sandwich ELISA kit in 96-well plate format.