Orders LinkContactUsLinkComponent
Ceramic D-Torch™
Ceramic D-Torch™
Assembly including quartz outer tube for OPTIMA™ ICP kit (8x00)
Catalog # PERCN0777840
Supplier:   PerkinElmer
Ceramic D-Torch™
Catalog # PERCN0777840
Supplier:  PerkinElmer
Ceramic D-Torch™
Assembly including quartz outer tube for OPTIMA™ ICP kit (8x00)
Price
1 120,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The ceramic outer tube is of particular benefit for the analysis of wear metals in engine oils, where quartz outer tubes often suffer from short lifetime.
  • Analysis of high-matrix solutions – Fusion samples, samples with high-dissolved solids, samples with high-acid content
  • Demountable quart or ceramic outer tubes
Also beneficial for Si determinations, where quartz outer tubes may produce high background signals.
Can cause early devitrification or sample deposition when using quartz.
Specifications
  • Description:
    Assembly including quartz outer tube for OPTIMA™ ICP kit (8x00)
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Ceramic outer tube
    Ceramic outer tube
    PERCN0777052
    OPTIMA™ ICP kits (4300/5300/7300V)
    Log in for availability
    1 * 1 ST - 2 140,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Quartz outer tube
    Quartz outer tube
    PERCN0777053
    OPTIMA™ ICP kits (4300/5300/7300V)
    Log in for availability
    1 * 1 ST - 283,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • DV D-Torch quartz including outer tube
    DV D-Torch quartz including outer tube
    PERCN0777508
    OPTIMA™ ICP kits (2x00/4x00/5x00/7x00)
    Log in for availability
    1 * 1 ST - 1 060,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • DV base and inner tube
    DV base and inner tube
    PERCN0777509
    OPTIMA™ ICP kits (2x00/4x00/5x00/7x00)
    Log in for availability
    1 * 1 ST - 700,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • DV quartz outer tube
    DV quartz outer tube
    PERCN0777510
    OPTIMA™ ICP kits (2x00/4x00/5x00/7x00)
    Log in for availability
    1 * 1 ST - 324,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Copper tape
    Copper tape
    PERCN0777843
    D-Torch outer tube
    Log in for availability
    1 * 12 ST - 53,60 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Quartz outer tube
    Quartz outer tube
    PERCN0777842
    OPTIMA™ ICP kit (8x00)
    Log in for availability
    1 * 1 ST - 337,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Torch adapter with auxiliary port
    Torch adapter with auxiliary port
    PERCN0774070
    OPTIMA™ ICP kits (4300/5300/7300V)
    Log in for availability
    1 * 1 ST - 741,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Ceramic outer tube
    Ceramic outer tube
    PERCN0777880
    OPTIMA™ ICP kit (8x00)
    Log in for availability
    1 * 1 ST - 2 960,00 €