Ammonia (nitrogen) test strips
Catalog # HACH2755325
Supplier: Hach
CAS Number:
Specifications
- Pk:25
- Description:Ammonia (nitrogen) test strips
- Environmentally Preferable:
- Tests per kit:25
- Range:0 - 6 mg/l
Specifications
About this item
Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish.
- Easy to use, disposable, and inexpensive
- A great way to obtain quick, quantitative answers in the field or in the lab
- Parameter: ammonia, nitrogen, low range - As NH₃-N, for freshwater