ContactUsLinkComponent
Accessories for temperature control units
Accessories for temperature control units
Catalog # JULA8980142
Supplier:  JULABO GmbH
undefined
Accessories for temperature control units
Catalog # JULA8980142
Supplier:  JULABO GmbH
Specifications
  • Description:
    Y-adapter cable for connecting 1 dual PT100 sensor to 2 Lemosa sockets for LC temperature laboratory controller
  • Pk:
    1