Order Entry
Belgium
ContactUsLinkComponent
 
Restriction enzymes, NotI, 50 U/µl, Fermentas
  ER0593
 :  
undefined
Restriction enzymes, NotI, 50 U/µl, Fermentas
  ER0593
 :  

 

  • Concentration:
    50 U/µl
  • Pk:
    1 KIT
  • Application:
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Source:
  • Description:
    NotI 50 U/µl
  • Seq:
    04
  • Environmentally Preferable:
  • Enzyme name:
    NotI
  • Units:
    1500 units

 

 

NotI restriction enzyme recognises GC^GGCCGC sites and cuts best at 37 °C in O buffer.

  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Applications: Molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP), SNP.