ContactUsLinkComponent
Silane adhesive slides, KF Frost
Silane adhesive slides, KF Frost
Catalog # 631-1238
Supplier:  KLINIPATH
Silane adhesive slides, KF Frost
Catalog # 631-1238
Supplier:  KLINIPATH
Specifications
  • Colour:
    White
  • Description:
    Silane adhesive slides with 90° ground edges
  • Pk:
    2.500
Specifications
About this item
These silane adhesive slides are specially designed for thermal transfer printers.
Certifications: CE certified according to IVDR (2017/746).