Wound closure system, Autoclip®
Supplier: HARVARD APPARATUS
Certificates
About this item
This Autoclip® wound closure system provides a rapid, easy to use method to securely close wounds. When the wound is healed, simply remove the clip using the handy removal forcep.
- Provides maximum holding force with minimal trauma
Automatic, spring-fed applicators permit rapid, unassisted application.
Delivery: Starter set includes 100 wound clips, applicator, and removal forcep.
Frequently Bought Together