ContactUsLinkComponent
Diffuser caps for waste and wash vials
Diffuser caps for waste and wash vials
Catalog # PERCN6101276
Supplier:  PerkinElmer
Diffuser caps for waste and wash vials
Catalog # PERCN6101276
Supplier:  PerkinElmer
Specifications
  • Description:
    Vial diffuser for waste and wash vials
  • Pk:
    1
Specifications
About this item
The conical shape allows the needle to penetrate in to solvent or waste.
  • Reduce spillage and large losses through evaporation