ContactUsLinkComponent
Manual pulse counter
Manual pulse counter
Catalog # MOIN1023MW
Supplier:  MOINEAU
undefined
Manual pulse counter
Catalog # MOIN1023MW
Supplier:  MOINEAU
Specifications
  • Range:
    0 − 9999
  • Description:
    Manual pulse counter
  • Material:
    Metal
  • Pk:
    1
Specifications
About this item
Manual mechanical counter with four digit key in a robust metal housing.
  • General reset by the rotary knob side
  • Adding one each finger pressure
  • Folding ring to attach it to your finger