Labels for BradyJet J2000 Lab printer
About this item
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
22 Options Available Below
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Product Family Options
Product Information
- TypeModelMaterialColourPrinting methodPrinter modelDimensionsPackedAvailabilityPrice
- BRDY150005Printer labelsPrinter labelsB-7425JJ20-152-7425JPPWhiteJ2000-Lab25×9 mm2000 labels/rollLog in for availability1 * 1 ROU151,19 €
- BRDY150007Printer labelsPrinter labelsB-7425JJ20-257-7425JPPWhiteJ2000-Lab25×9 mm1500 labels/rollLog in for availability1 * 1 ROU180,39 €
- BRDY150023Labels with 9 mm dotLabels with 9 mm dotB-7425JJ20-249-7425JPPWhiteJ2000-Lab25×9 mm1500 labels/rollLog in for availability1 * 1 ROU186,00 €
- BRDY150006Printer labelsPrinter labelsB-7425JJ20-59-7425JPPWhiteJ2000-Lab25×12 mm2000 labels/rollLog in for availability1 * 1 ROU164,60 €
- BRDY150033Labels with 11 mm dotLabels with 11 mm dotB-7425JJ20-251-7425JPPWhiteJ2000-Lab25×12 mm1500 labels/rollLog in for availability1 * 1 ROU178,47 €
- BRDY150015Printer labelsPrinter labelsB-7425JJ20-88-7425JPPWhiteJ2000-Lab25×19 mm2000 labels/rollLog in for availability1 * 1 ROU321,33 €
- BRDY150025Printer labelsPrinter labelsB-7425JJ20-179-7425JPPWhiteJ2000-Lab25×25 mm1500 labels/rollLog in for availability1 * 1 ROU162,02 €
- BRDY150542Label printer tapeLabel printer tapeB-7425JJ20C-1125-7425JPPWhiteThermal transferJ2000-Lab29 mm×30 m1 roll/boxLog in for availability1 * 1 ST175,00 €
- BRDY150016Printer labelsPrinter labelsB-7425JJ20-256-7425JPPWhiteJ2000-Lab33×15 mm1500 labels/rollLog in for availability1 * 1 ROU147,81 €
- BRDY150020Printer labelsPrinter labelsB-7425JJ20-37-7425JPPWhiteJ2000-Lab38×12 mm1500 labels/rollLog in for availability1 * 1 ROU157,54 €
- BRDY150019Printer labelsPrinter labelsB-7425JJ20-136-7425JPPWhiteJ2000-Lab38×19 mm1500 labels/rollLog in for availability1 * 1 ROU228,61 €
- BRDY150022Printer labelsPrinter labelsB-7425JJ20-235-7425JPPWhiteJ2000-Lab38×25 mm1250 labels/rollLog in for availability1 * 1 ROU97,80 €
- BRDY150028Printer labelsPrinter labelsB-7425JJ20-258-7425JPPWhiteJ2000-Lab38×44 mm900 labels/rollLog in for availability1 * 1 ROU155,70 €
- BRDY150011Printer labelsPrinter labelsB-7425JJ20-151-7425JPPWhiteJ2000-Lab50×6 mm2000 labels/rollLog in for availability1 * 1 ROU149,44 €
- BRDY150024Printer labelsPrinter labelsB-7425JJ20-53-7425JPPWhiteJ2000-Lab50×12 mm2000 labels/rollLog in for availability1 * 1 ROU216,03 €
- BRDY150018Printer labelsPrinter labelsB-7425JJ20-137-7425JPPWhiteJ2000-Lab50×25 mm1500 labels/rollLog in for availability1 * 1 ROU123,25 €
- BRDY150543Printer labelsPrinter labelsB-7425JJ20-260-7425JPPWhiteJ2000-Lab55×85 mm340 labels/rollLog in for availability1 * 340 ST263,00 €
- BRDY150547Label printer tapeLabel printer tapeB-7425JJ20C-2250-7425JPPWhiteThermal transferJ2000-Lab57 mm×30 m1 roll/boxLog in for availability1 * 1 ST259,00 €
- BRDY150541Printer labelsPrinter labelsB-7425JJ20-261-7425JPPWhiteJ2000-Lab75×110 mm270 labels/rollLog in for availability1 * 270 ST290,00 €
- BRDY150545Printer labelsPrinter labelsB-7425JJ20-18-7425JPPWhiteJ2000-Lab76×25 mm1000 labels/rollLog in for availability1 * 1 ROU339,00 €
- BRDY150544Printer labelsPrinter labelsB-7425JJ20-262-7425JPPWhiteJ2000-Lab105×155 mm200 labels/rollLog in for availability1 * 200 ST400,00 €
Ink pads
Product Information
- TypeModelMaterialColourPrinting methodPrinter modelDimensionsPackedAvailabilityPrice
- BRDY150163Ink pads