Orders LinkContactUsLinkComponent
Label printer, colour, J2000
Label printer, colour, J2000
Catalog #
Label printer, colour, J2000
Catalog #
Supplier:  
Label printer, colour, J2000
Label printer, colour, J2000
Product Details & Documents
The BradyJet J2000 is a practical and easy to use, full colour inkjet label printer that can print reliable, high resolution signs, facility and laboratory sample labels. Suitable for many applications including lean/5S applications and die-cut materials for wire, laboratory or general identification, GHS / CLP labels, pipe markers, equipment labels, inventory identification, tags, tube labels, cryo labels and glassware labels.
  • High, photo quality printing resolution of 4800 dpi
  • Quality images, barcodes and pictures can be added to any sign
  • Easy colour coding as can print in any colour
  • Print speed of 63 mm/second enables larger colour coded labels to be printed on demand quickly