Order Entry
Belgium
ContactUsLinkComponent
Acrylamide standard, 1000 μg/ml in methanol
Acrylamide standard, 1000 μg/ml in methanol
Catalog # REST30494
Supplier:  Restek
CAS Number:  
Acrylamide standard, 1000 μg/ml in methanol
Catalog # REST30494
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 mL
  • CAS No.:
    79-06-1
  • Description:
    Acrylamide, 1000 µg/ml in methanol
  • Formula:
    C₃H₅NO
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.