ContactUsLinkComponent
8260B acetate mix
8260B acetate mix
  REST30489
 :  Restek
 :  
8260B acetate mix
  REST30489
 :  Restek
 :  
  • Concentration:
    2000 μg/ml each
  • Pk:
    1 mL
  • Description:
    8260B acetate mix, 7 components
  • Pk Info:
    1 ml/ampoule
  • Type:
    US EPA 8260B
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
This mix includes methyl acetate and n-amyl acetate. Contains seven acetates, 2000 µg/ml each in P and T methanol
Contents: n-amyl acetate, butyl acetate, ethyl acetate, isopropyl acetate, methyl acetate, propyl acetate and vinyl acetate.