531.1 Performance Check Mix
Catalog # REST32275
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:531.1 Performance check mix in methanol
- Pk Info:1 ml/ampoule
Specifications
About this item
US EPA 531.1 check mix
This mix contains four components.
Components: Aldicarb sulphoxide (100 µg/ml), BDMC (10 µg/ml), 3-hydroxycarbofuran (2 µg/ml), Methiocarb (20 µg/ml).