DNPH reference materials
About this item
The reported concentrations reflect the amount of aldehyde or ketone in the mixture.The concentration of derivatised aldehyde or ketone is not reported.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
3 Options Available Below
Product Details & Documents
The reported concentrations reflect the amount of aldehyde or ketone in the mixture.The concentration of derivatised aldehyde or ketone is not reported.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Product Family Options
Product Information
- ConcentrationMatrixPk InfoPkAvailabilityPrice
- REST33074Acetaldehyde-2,4-DNPH
- REST33082Formaldehyde-2,4-DNPH
- REST33091Glycolaldehyde-2,4-DNPHGlycolaldehyde-2,4-DNPH100 µg/mlCH₃CN1 ml/ampoule1 AmpoulLog in for availability1 * 1 Ampul61,30 €
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.