Herbicide Surrogate (2,4-DCAA)
Supplier: Restek
Certificates
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
- CAS No.:19719-28-9
- Formula:Cl₂C₆H₃CH₂CO₂H
- Description:2,4-Dichlorophenylacetic acid
Specifications
Frequently Bought Together