ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å), beads 8 - 12 mesh, Fluka™
Molecular sieve A4 (0.4 nm, 4 Å), beads 8 - 12 mesh, Fluka™
  • Particle Size:
    8-12 mesh
  • Grain size 8 - 12 mesh:
    Min. 95 %
Frequently Bought Together