Order Entry
Belgium
ContactUsLinkComponent
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Flammable
Health
Irritant
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293253-1L
CAS Number:  
undefined
Flammable
Health
Irritant
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293253-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Autoignition Temperature:
    437 °F
  • Identity (IR):
    complying
  • Assay of hexane (all isomers, GC):
    Min. 98.50 %
  • Refractive index (n 20/D):
    1.377 - 1.382
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Titrable Acid:
    Max. 0.0003 meq/g
  • S-compounds (as S):
    Max. 0.005 %
  • Thiophene:
    complying
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 250 nm:
    Max. 0.01
  • Absorbance at 240 nm:
    Max. 0.02
  • Absorbance at 230 nm:
    Max. 0.05
  • Absorbance at 220 nm:
    Max. 0.10
  • Absorbance at 210 nm:
    Max. 0.30
  • Absorbance at 195 nm:
    Max. 1.0
  • APHA:
    Max. 10
  • Vapor Density:
    3 (vs air)
  • Explosion Limit:
    7.5 %

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:48…65 °C (1.013 hPa)
Melting Pt:–154 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–26 °C
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311