Order Entry
Belgium
ContactUsLinkComponent
Sodium formate ≥99.0% ACS, Fluka™
Sodium formate ≥99.0% ACS, Fluka™
Catalog # 247596-500G
CAS Number:  
Sodium formate ≥99.0% ACS, Fluka™
Catalog # 247596-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay:
    Min. 99.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.001 %

Specification Test Results

About this item

 :
 :68.01 g/mol
Merck Index:13,08694
Melting Pt:255 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-488-0
REACH:01-2119486468-21
CAS Number:141-53-7
Formula:HCO₂Na
MDL Number:MFCD00013101