Order Entry
Belgium
ContactUsLinkComponent
Manganese(II) chloride tetrahydrate ≥99%, Puriss. p.a. ACS, Fluka™
Corrosive
Health
Toxic
Manganese(II) chloride tetrahydrate ≥99%, Puriss. p.a. ACS, Fluka™
Certificates
Corrosive
Health
Toxic
Manganese(II) chloride tetrahydrate ≥99%, Puriss. p.a. ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • pH (5 %, 25 °C):
    3.5 - 6.0
  • Calcium (Ca):
    max. 0.001 %
  • Copper (Cu):
    max. 0.0005 %
  • Iron (Fe):
    max. 5 ppm
  • Potassium (K):
    max. 0.001 %
  • Lead (Pb):
    max. 0.0005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sulfate (SO₄):
    max. 0.005 %
  • Sodium (Na):
    Max. 0.005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • KMnO₄ reducing matter (as O):
    Max. 0.0005 %
  • Water insoluble matter:
    Max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together