Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate, anhydrous ≥99.5%, Reagent Grade
Magnesium sulphate, anhydrous ≥99.5%, Reagent Grade
Certificates
undefined
Magnesium sulphate, anhydrous ≥99.5%, Reagent Grade

Specification Test Results

  • Assay (calculated to the dried substance):
    Min. 99.5 %
  • Loss on drying (105°C):
    Max. 2 %
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together