Order Entry
Belgium
ContactUsLinkComponent
 
Sodium acetate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
 
Sodium acetate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
 

 

  • Assay (HClO₄-titr.):
    99.0 - 101.0 %
  • Absorbance at 250 nm:
    Max. 0.05
  • Absorbance at 260 nm:
    Max. 0.01
  • Absorbance 500 nm:
    Max. 0.01
  • Appearance of the solution (c=10%):
    complying
  • pKa :
    4.76 (acetic acid)
  • Form:
    Solid
  • Autoignition Temperature:
    1112 °F

 

 

Frequently Bought Together