Order Entry
Belgium
ContactUsLinkComponent
Bromothymol blue solution in ethanol acid-base indicator, Fluka™
Irritant
Flammable
Bromothymol blue solution in ethanol acid-base indicator, Fluka™
Catalog # 32822-100ML
CAS Number:  
Irritant
Flammable
Bromothymol blue solution in ethanol acid-base indicator, Fluka™
Catalog # 32822-100ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Test of sensitivity:
    complying
  • Change range (pH 5,8-7,4):
    complying

Specification Test Results

About this item

 :
Flash Pt:38 °C
Storage Temperature:Ambient
UN:1170
ADR:3,III
 :624.39 g/mol
 :GHS07
 :GHS02
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872