Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Certificates
Sodium hypochlorite ∼10% (RT) in aqueous solution, Purum, Fluka™
Certificates

Specification Test Results

  • Assay of active chlorine:
    8.0 - 15.0 %
  • pH (direct):
    11.5 - 13.5
  • Density (D 20/4):
    1.21 - 1.26

Specification Test Results

Related Information

Frequently Bought Together