Diammonium hydrogen orthophosphate 99-102%, Puriss., meets analytical specification of NF, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Merck Index:13,00546
Melting Pt:155 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-987-8
REACH:01-2119490974-22
Specification Test Results
- Assay:98.0 - 102.0 %
- pH (1 %,25°C):7.6 - 8.2
- Arsenic (As):Max. 2 ppm
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.002 %
- Sulfate (SO₄):Max. 0.02 %
Specification Test Results
Frequently Bought Together