Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Irritant
Flammable
tert-Butyl methyl ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Certificates
undefined
Irritant
Flammable
tert-Butyl methyl ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Density (20°C):
    0.736 - 0.744 g/ml
  • Refractive index (n 20/D):
    1.362 - 1.376
  • Non-volatile matter:
    Max. 0.005 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Free acid (as CH₃COOH):
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Peroxides (as H₂O₂):
    Max. 0.0005 %
  • Appearance (color):
    complying
  • Appearance (form):
    complying
  • Autoignition Temperature:
    705 °F

Specification Test Results

Related Information

Frequently Bought Together