Order Entry
Belgium
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Certificates
Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Certificates

Specification Test Results

  • Assay (as MoO₃):
    81.0 - 83.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.01 %
  • Heavy metals (ICP) ACS:
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.002 %
  • Nitrate (NO₃):
    Max. 0.003 %
  • Phosphate, Arsenate, Silicate (as SiO₂):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.02 %

Specification Test Results

Related Information

Frequently Bought Together