
Ammonium hydrogen carbonate ≥99% eluent additive, for LC-MS, Fluka™
Catalog # 40867-50G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Assay:99.5 - 101.0 %
- pH (1 M, 20°C):7.0 - 8.5
- Solution 1 M in H₂O:complying
- Aluminium (Al):Max. 5 ppm
- Barium (Ba):Max. 5 ppm
- Bismuth (Bi):Max. 5 ppm
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Lithium (Li):Max. 5 ppm
- Magnesium (Mg):Max. 500 ppm
- Manganese (Mn):Max. 5 ppm
- Molybdenum (Mo):Max. 5 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Strontium (Sr):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 50 ppm
- Total S (as SO₄):Max. 50 ppm
- Suitability for the LC-MS:complying
Specification Test Results
About this item
:
:79.06 g/mol
Merck Index:13,00497
Melting Pt:60 °C
Density:1.58 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
Storage Temperature:Ambient
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138