Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.1 M (0.1 N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide 0.1 M (0.1 N) volumetric solution, Fluka™
Catalog # 71395-1L
CAS Number:  
Corrosive
Sodium hydroxide 0.1 M (0.1 N) volumetric solution, Fluka™
Catalog # 71395-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    0.0998 - 0.1002 mol/l
  • Calibrated Against:
    potassium hydrogen phthalate

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548