Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™
Corrosive
Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™
Certificates
Corrosive
Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™
Certificates

Specification Test Results

  • Assay:
    98.0 - 100.5 %
  • Assay of Na₂CO₃:
    Max. 0.5 %
  • Aluminium (Al):
    Max. 0.001 %
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 0.001 %
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Potassium (K):
    Max. 0.1 %
  • Lead (Pb):
    Max. 0.5 ppm
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Chloride (Cl):
    Max. 100 ppm
  • Total N:
    Max. 0.0005 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Silicate (as SiO₂):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 100 ppm
  • Water insoluble matter + org. comp.:
    complying
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying
  • Vapor Pressure:
    <18 mmHg ( 20 °C)

Specification Test Results

Related Information

Frequently Bought Together