Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide ~50% in aqueous solution for HPLC, Fluka™
Corrosive
Sodium hydroxide ~50% in aqueous solution for HPLC, Fluka™
Certificates
Corrosive
Sodium hydroxide ~50% in aqueous solution for HPLC, Fluka™
Certificates

Specification Test Results

  • Assay:
    49.0 - 51.0 %
  • Density (D 20/4):
    1.500 - 1.540
  • Carbonate (as Na₂CO₃):
    Max. 0.1 %
  • Absorbance 500 nm:
    Max. 0.02
  • Absorbance at 300 nm:
    Max. 0.03
  • Absorbance 280 nm:
    Max. 0.08
  • Absorbance at 260 nm:
    Max. 0.15
  • Absorbance at 240 nm:
    Max. 0.17
  • Suitability test (HPLC):
    complying

Specification Test Results

Related Information

Frequently Bought Together