Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Health
Potassium iodide 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Certificates
Health
Potassium iodide 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Certificates

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (105°C):
    Max. 0.5 %
  • Alkalinely reacting substances:
    complying
  • Iron (Fe):
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Iodate (JO₃):
    complying
  • Iodate (IO₃):
    Max. 0.0004 %
  • Sulfate (SO₄):
    Max. 100 ppm
  • Thiosulfate (S₂O₃):
    Max. 0.01 %
  • Nitrate, nitrite, ammonium:
    complying
  • Thiosulfate (S₂O₃) and Ba:
    complying
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together