Sodium chloride ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1.465 °C (1.013 hPa)
Melting Pt:800 °C
Density:2.160 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Assay:Min. 99.5 %
- pH (5 %, 20°C):5.0 - 8.0
- Arsenic (As):Max. 0.00001 %
- Barium (Ba):Max. 0.0005 %
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0001 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.0005 %
- Manganese (Mn):Max. 0.0005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Bromide (Br):Max. 0.005 %
- Iodide (I):Max. 0.001 %
- Potassium hexacyanoferrate(II):Max. 0.0001 %
- Phosphate (PO₄):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.001 %
- Total N:Max. 0.001 %
- Appearance of the solution:complying
Specification Test Results
Frequently Bought Together