Order Entry
Belgium
ContactUsLinkComponent
Potassium bromide ≥99% (trace metals basis), FT-IR Grade, Fluka™
Potassium bromide ≥99% (trace metals basis), FT-IR Grade, Fluka™
Certificates
Potassium bromide ≥99% (trace metals basis), FT-IR Grade, Fluka™
Certificates

Specification Test Results

  • Suitability for IR spectroscopy:
    complying
  • Assay:
    Min. 99 %
  • Metallic impurities (Total):
    Max. 10000 ppm
  • Vapor Pressure:
    <0.01 mmHg ( 20 °C)

Specification Test Results

Related Information

Frequently Bought Together