


Cyclohexane ≥99% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1.013 hPa)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS02
:GHS08
:GHS07
Specification Test Results
- Vapor Pressure:168.8 mmHg ( 37.7 °C)
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.02 %
- Reaction against H₂SO₄:complying
- UV-absorption:complying
- APHA:Max. 10
- Refractive Index:n20/D 1.426 (lit.)
Specification Test Results
Frequently Bought Together