o-Xylene 98%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 295884-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 97.5 %
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.02 %
- Identity (IR):complying
- Absorbance at 350 nm:Max. 0.01
- Absorbance at 325 nm:Max. 0.05
- Absorbance at 300 nm:Max. 0.40
- Absorbance at 288 nm:Max. 1.00
- Autoignition Temperature:867 °F
- Vapor Pressure:16 mmHg ( 37.7 °C)
Specification Test Results
About this item
Synonyms: 1,2-Dimethylbenzene
:
:106.17 g/mol
Flash Pt:7.6 %(V)
EINECS:202-422-2
UN:1307
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:95-47-6
Formula:C₈H₁₀
MDL Number:MFCD00008519
Boiling Pt:143…145 °C (1.013 hPa)
Melting Pt:–25 °C
Density:0.88 g/cm³ (20 °C)
ADR:3 III
Merck Index:12,10214