

n-Decane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:124-18-5
:
Formula:H₃C(CH₂)₈CH₃
MDL Number:MFCD00008954
:142.28 g/mol
Boiling Pt:2.6 %(V)
Melting Pt:–27 °C
Density:0.732 g/cm³ (20 °C)
Flash Pt:57.4 °C (1.013.25 hPa)
Storage Temperature:Ambient
EINECS:204-686-4
UN:2247
ADR:3,III
:GHS08
:GHS02
Specification Test Results
- Vapor Pressure:3.77 mmHg ( 37.7 °C)
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.409 - 1.413
- Explosion Limit:2.6 %
- Autoignition Temperature:410 °F
- Vapor Density:4.9 (vs air)
Specification Test Results
Frequently Bought Together