Order Entry
Belgium
ContactUsLinkComponent
 
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
    
 
undefined
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

 

 

  • Assay (GC):
    Min. 99.5 %
  • Density (D 20/4):
    1.261 - 1.263
  • Refractive index (n 20/D):
    1.473 - 1.475
  • Water (Karl Fischer):
    Max. 0.1 %
  • Residue on ignition:
    Max. 0.005 %
  • Indifference:
    complying
  • Calcium (Ca):
    Max. 0.0005 %
  • Cadmium (Cd):
    Max. 0.0001 %
  • Cobalt (Co):
    Max. 0.0001 %
  • Chromium (Cr):
    Max. 0.0001 %
  • Copper (Cu):
    Max. 0.0001 %
  • Iron (Fe):
    Max. 0.0001 %
  • Potassium (K):
    Max. 0.002 %
  • Magnesium (Mg):
    Max. 0.0001 %
  • Manganese (Mn):
    Max. 0.0001 %
  • Sodium (Na):
    Max. 0.002 %
  • Nickel (Ni):
    Max. 0.0001 %
  • Lead (Pb):
    Max. 0.0001 %
  • Zinc (Zn):
    Max. 0.0001 %
  • Heavy metals (as Pb):
    Max. 0.0002 %
  • Halogene compounds (as Cl):
    Max. 0.003 %
  • Ammonium (NH₄):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.0001 %
  • Sulfate (SO₄):
    Max. 0.001 %
  • Reaction against H₂SO₄:
    complying
  • Acrolein, Glucose + NH4-compounds:
    complying
  • Fatty acids and ester (as butyric acid):
    Max. 0.05 %
  • APHA:
    Max. 10

 

 

Frequently Bought Together