Order Entry
Belgium
ContactUsLinkComponent
Methanol ≥99.93%, BioReagent for protein sequencing, Burdick & Jackson™
Flammable
Toxic
Health
Methanol ≥99.93%, BioReagent for protein sequencing, Burdick & Jackson™
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.93%, BioReagent for protein sequencing, Burdick & Jackson™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.925 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Titrable Acid:
    Max. 0.0003 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 260 nm:
    Max. 0.01
  • Absorbance at 240 nm:
    Max. 0.05
  • Absorbance at 230 nm:
    Max. 0.15
  • Absorbance at 220 nm:
    Max. 0.30
  • Absorbance at 210 nm:
    Max. 0.60
  • Absorbance at 205 nm:
    Max. 1.00
  • Application test:
    complying
  • APHA:
    Max. 10

Specification Test Results

Related Information

Frequently Bought Together