Order Entry
Belgium
ContactUsLinkComponent
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Flammable
Corrosive
Irritant
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
   Sustainable Options Available
Certificates
undefined
Flammable
Corrosive
Irritant
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    116 - 118 °C
  • Refractive Index:
    n20/D 1.399 (lit.)
  • Density (D 20/20):
    0.808 - 0.812
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as C₃H₇COOH):
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • iso-butanol (GC):
    Max. 0.15 %
  • n-Butyraldehyde (GC):
    Max. 0.01 %
  • Di-n-butyl ether (GC):
    Max. 0.1 %
  • 2-Butanol (GC):
    Max. 0.05 %
  • Carbonyl compounds (as CO):
    Max. 0.01 %
  • APHA:
    Max. 10

Specification Test Results

Related Information

Frequently Bought Together