

Trifluoroacetic acid 99%, Reagent Grade, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:72 °C (1.013 hPa)
Melting Pt:–15 °C
Density:1.48 g/cm³ (20 °C)
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
Specification Test Results
- Vapor Pressure:97.5 mmHg ( 20 °C)
- Assay (alkalimetric):98.5 - 101.5 %
- Assay (GC):Min. 98.5 %
- Identity (IR):complying
- APHA:Max. 30
- Refractive Index:n20/D 1.3 (lit.)
Specification Test Results
Frequently Bought Together