Order Entry
Belgium
ContactUsLinkComponent
Ammonium acetate ≥99.0%, CHROMASOLV™ LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Ammonium acetate ≥99.0%, CHROMASOLV™ LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Catalog # 14267-25G
CAS Number:  
undefined
Ammonium acetate ≥99.0%, CHROMASOLV™ LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Catalog # 14267-25G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • Water (Karl Fischer):
    Max. 2.0 %
  • Residue on ignition:
    Max. 0.01 %
  • pH (1 M, 20°C):
    6.2 - 7.5
  • Solution 1 M in H₂O:
    complying
  • Aluminium (Al):
    Max. 1 ppm
  • Arsenic (As):
    Max. 0.1 ppm
  • Barium (Ba):
    Max. 1 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 5 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Cobalt (Co):
    Max. 1 ppm
  • Chromium (Cr):
    Max. 1 ppm
  • Copper (Cu):
    Max. 1 ppm
  • Iron (Fe):
    Max. 1 ppm
  • Potassium (K):
    Max. 5 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 1 ppm
  • Manganese (Mn):
    Max. 1 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 1 ppm
  • Strontium (Sr):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 1 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Nitrate (NO₃):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Suitability for the UHPLC-MS:
    complying

Specification Test Results

About this item

 :
 :77.08 g/mol
Merck Index:13,00495
Melting Pt:113 °C
Density:1.170 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066